Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021216.4 | NP_067039.1 | 3 | 558 | ||
XM_011527193.2 | XP_011525495.1 | 4 | 710 | |||
XM_017027078.1 | XP_016882567.1 | 4 | 620 | |||
XM_017027079.1 | XP_016882568.1 | 4 | 2490 | |||
XM_017027080.1 | XP_016882569.1 | 3 | 493 | |||
GenBank mRNA | AF269249.1 | - | 2 | 404 | ||
BC014280.2 | - | 3 | 530 | 3840512 |