Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM60A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135811.1 | NP_001129283.1 | 3 | 408 | ||
NM_001135812.1 | NP_001129284.1 | 2 | 304 | |||
NM_021238.2 | NP_067061.1 | 3 | 404 | |||
XM_011520803.2 | XP_011519105.1 | 2 | 406 | |||
XM_017019758.1 | XP_016875247.1 | 3 | 472 | |||
XM_017019759.1 | XP_016875248.1 | 4 | 518 | |||
XM_017019760.1 | XP_016875249.1 | 4 | 523 | |||
XM_017019761.1 | XP_016875250.1 | 3 | 453 | |||
XM_017019762.1 | XP_016875251.1 | 3 | 370 | |||
GenBank mRNA | AF087885.1 | - | 3 | 357 | ||
AF212220.1 | - | 2 | 263 | |||
AK026932.1 | - | 2 | 215 | |||
AK294457.1 | - | 2 | 317 | |||
AL157432.1 | - | 3 | 356 | |||
AY598323.1 | - | 1 | 82 | |||
BC000024.2 | - | 3 | 404 | 3504430 | ||
BC071966.1 | - | 2 | 184 | 5924274 | ||
BX648630.1 | - | 1 | 399 | |||
DA741767.1 | - | 2 | 451 |