Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WIZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021241.2 | NP_067064.2 | 6, 7 | 2232 | ||
XM_005260004.4 | XP_005260061.1 | 11, 12 | 5467 | |||
XM_005260005.4 | XP_005260062.1 | 10, 11 | 5296 | |||
XM_005260006.4 | XP_005260063.1 | 10, 11 | 5068 | |||
XM_005260007.4 | XP_005260064.1 | 9, 10 | 4897 | |||
XM_005260008.2 | XP_005260065.1 | 9, 10 | 3311 | |||
XM_005260009.2 | XP_005260066.1 | 7, 8 | 2658 | |||
XM_005260010.3 | XP_005260067.1 | 7, 8 | 2671 | |||
XM_005260011.2 | XP_005260068.1 | 7, 8 | 2741 | |||
XM_005260012.2 | XP_005260069.1 | 6, 7 | 2259 | |||
XM_006722828.3 | XP_006722891.1 | 9, 10 | 3080 | |||
XM_011528163.2 | XP_011526465.1 | 7, 8 | 2732 | |||
GenBank mRNA | AK027615.1 | - | 3, 4 | 1186 | ||
AK091183.1 | - | 4, 5 | 1692 | |||
AK122890.1 | - | 7, 8 | 2288 | |||
AK131404.1 | - | 6, 7 | 2233 | |||
AL390184.1 | - | 1, 2 | 104 | |||
BC002329.2 | - | 1, 2 | 95 | |||
BC007551.1 | - | 2, 3 | 744 | |||
BC062360.1 | - | 3, 4 | 956 | |||
BC098445.1 | - | 3, 4 | 1466 | |||
BC136641.1 | - | 5, 6 | 2165 | 9020631 | ||
BC144332.1 | - | 6, 7 | 2564 | 9052852 |