Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004161.4 | NP_004152.1 | 5 | 768 | ||
XM_005264468.1 | XP_005264525.1 | 4 | 477 | |||
GenBank mRNA | AF498929.1 | - | 5 | 382 | ||
AK055927.1 | - | 3 | 383 | |||
AK303678.1 | - | 4 | 476 | |||
AK315495.1 | - | 5 | 577 | |||
BC000905.2 | - | 5 | 560 | 2900705 | ||
BT019528.1 | - | 5 | 382 | |||
BT019529.1 | - | 5 | 382 | |||
BX571747.1 | - | 5 | 565 | |||
CR536488.1 | - | 5 | 382 | |||
M28209.1 | - | 5 | 432 | |||
X13898.1 | - | 5 | 382 |