Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243738.1 | NP_001230667.1 | 16 | 2403 | ||
NM_004761.4 | NP_004752.1 | 17 | 2600 | |||
XM_011514781.1 | XP_011513083.1 | 14 | 1797 | |||
GenBank mRNA | AB012295.1 | - | 15 | 2031 | ||
AK095967.1 | - | 14 | 1574 | |||
AL050259.1 | - | 17 | 2173 | |||
BC000316.1 | - | 11 | 1259 | |||
BC005037.1 | - | 14 | 1641 | |||
BC032681.1 | - | 17 | 2155 | 5585507 | ||
BC040989.1 | - | 16 | 2455 | |||
BX648015.1 | - | 16 | 1945 | |||
D85757.1 | - | 2 | 102 | |||
U68142.1 | - | 11 | 1209 |