Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006908.4 | NP_008839.2 | 5, 6 | 683 | ||
NM_018890.3 | NP_061485.1 | 6, 7 | 740 | |||
GenBank mRNA | AB451238.1 | - | 5, 6 | 442 | ||
AB451363.1 | - | 5, 6 | 442 | |||
AF136373.1 | - | 6, 7 | 499 | |||
AF498964.1 | - | 5, 6 | 442 | |||
AJ132694.1 | - | 6, 7 | 499 | |||
AK054993.1 | - | 5, 6 | 658 | |||
AK291806.1 | - | 5, 6 | 639 | |||
AK291815.1 | - | 6, 7 | 686 | |||
AY279384.1 | - | 5, 6 | 600 | |||
BC004247.1 | - | 5, 6 | 580 | 3610745 | ||
BC050687.1 | - | 5, 6 | 630 | 6149377 | ||
BC107748.1 | - | 5, 6 | 645 | 6699945 | ||
BQ689587.1 | - | 6, 7 | 699 | |||
BT007121.1 | - | 5, 6 | 442 | |||
M29870.1 | - | 5, 6 | 442 | |||
M31467.1 | - | 5, 6 | 442 |