Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278622.1 | NP_001265551.1 | 14 | 1841 | ||
NM_002873.1 | NP_002864.1 | 13 | 1946 | |||
NM_133338.2 | NP_579916.1 | 15 | 1842 | |||
NM_133339.2 | NP_579917.1 | 12 | 1399 | |||
NM_133340.2 | NP_579918.1 | 12 | 1432 | |||
NM_133341.2 | NP_579919.1 | 12 | 1358 | |||
NM_133342.2 | NP_579920.1 | 14 | 1819 | |||
NM_133343.1 | NP_579921.1 | 14 | 1784 | |||
NM_133344.2 | NP_579922.1 | 13 | 1601 | |||
XM_017009679.1 | XP_016865168.1 | 14 | 1661 | |||
XM_017009680.1 | XP_016865169.1 | 13 | 1715 | |||
XM_017009681.1 | XP_016865170.1 | 13 | 1573 | |||
GenBank mRNA | AF017748.1 | - | 13 | 1571 | ||
AF076838.1 | - | 13 | 1946 | |||
AF098533.1 | - | 12 | 1378 | |||
AF098534.1 | - | 12 | 1304 | |||
AF112263.1 | - | 12 | 1423 | |||
AF126424.1 | - | 15 | 1841 | |||
AJ001642.1 | - | 13 | 1610 | |||
AJ004977.1 | - | 13 | 1606 | |||
AK292487.1 | - | 14 | 1819 | |||
AL122068.1 | - | 14 | 1784 | |||
BC032304.1 | - | 12 | 1412 | 5170876 | ||
BX537441.1 | - | 14 | 1841 |