Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278622.1 | NP_001265551.1 | 9 | 1290 | ||
NM_002873.1 | NP_002864.1 | 8 | 1395 | |||
NM_133338.2 | NP_579916.1 | 10 | 1291 | |||
NM_133339.2 | NP_579917.1 | 7 | 848 | |||
NM_133340.2 | NP_579918.1 | 7 | 881 | |||
NM_133341.2 | NP_579919.1 | 7 | 807 | |||
NM_133342.2 | NP_579920.1 | 9 | 1268 | |||
NM_133343.1 | NP_579921.1 | 9 | 1233 | |||
NM_133344.2 | NP_579922.1 | 8 | 1050 | |||
XM_017009679.1 | XP_016865168.1 | 9 | 1110 | |||
XM_017009680.1 | XP_016865169.1 | 8 | 1164 | |||
XM_017009681.1 | XP_016865170.1 | 8 | 1022 | |||
GenBank mRNA | AF017748.1 | - | 8 | 1020 | ||
AF076838.1 | - | 8 | 1395 | |||
AF085736.2 | - | 7 | 848 | |||
AF098533.1 | - | 7 | 827 | |||
AF098534.1 | - | 7 | 753 | |||
AF112263.1 | - | 7 | 872 | |||
AF126424.1 | - | 10 | 1290 | |||
AJ001642.1 | - | 8 | 1059 | |||
AJ004977.1 | - | 8 | 1055 | |||
AK292487.1 | - | 9 | 1268 | |||
AL122068.1 | - | 9 | 1233 | |||
BC032304.1 | - | 7 | 861 | 5170876 | ||
BX537441.1 | - | 9 | 1290 |