Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042368.2 | NP_001035827.1 | 15 | 2267 | ||
NM_001271774.1 | NP_001258703.1 | 15 | 2179 | |||
NM_001271775.1 | NP_001258704.1 | 15 | 2272 | |||
NM_001271776.1 | NP_001258705.1 | 15 | 2252 | |||
NM_006266.3 | NP_006257.1 | 15 | 2288 | |||
GenBank mRNA | AB037729.1 | - | 11 | 1580 | ||
AF027169.1 | - | 15 | 2183 | |||
AF295773.1 | - | 15 | 2203 | |||
AK000242.1 | - | 10 | 1245 | |||
AK024575.1 | - | 7 | 842 | |||
AK074114.1 | - | 15 | 2158 | |||
AK090450.1 | - | 11 | 1614 | |||
AK127524.1 | - | 15 | 2272 | |||
AK301470.1 | - | 15 | 2139 | |||
BC021581.1 | - | 3 | 364 | |||
BC033198.1 | - | 7 | 684 | |||
BC059362.1 | - | 15 | 2242 | 30340922 | ||
U14417.1 | - | 4 | 434 |