Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RALGDS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042368.2 | NP_001035827.1 | 17 | 2545 | ||
NM_001271774.1 | NP_001258703.1 | 17 | 2457 | |||
NM_001271775.1 | NP_001258704.1 | 17 | 2550 | |||
NM_001271776.1 | NP_001258705.1 | 17 | 2530 | |||
NM_006266.3 | NP_006257.1 | 17 | 2566 | |||
GenBank mRNA | AB037729.1 | - | 11 | 3170 | ||
AF027169.1 | - | 17 | 2461 | |||
AF295773.1 | - | 17 | 2481 | |||
AK000242.1 | - | 12 | 1523 | |||
AK024575.1 | - | 9 | 1120 | |||
AK056462.1 | - | 16 | 2243 | |||
AK090450.1 | - | 12 | 3029 | |||
AK127524.1 | - | 17 | 2550 | |||
AK301470.1 | - | 17 | 2417 | |||
BC021581.1 | - | 5 | 642 | |||
BC033198.1 | - | 9 | 962 | |||
BC059362.1 | - | 17 | 2520 | 30340922 | ||
BM805052.1 | - | 2 | 131 | |||
U14417.1 | - | 6 | 712 |