Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303051.1 | NP_001289980.1 | 2 | 262 | ||
NM_001303109.1 | NP_001290038.1 | 2 | 282 | |||
NM_021633.3 | NP_067646.1 | 2 | 282 | |||
XM_011509835.1 | XP_011508137.1 | 3 | 302 | |||
XM_011509836.1 | XP_011508138.1 | 3 | 302 | |||
XM_011509837.1 | XP_011508139.1 | 3 | 302 | |||
XM_017001995.1 | XP_016857484.1 | 2 | 251 | |||
GenBank mRNA | AF190900.1 | - | 2 | 206 | ||
AF306689.1 | - | 2 | 251 | |||
AK027656.1 | - | 2 | 253 | |||
AK223072.1 | - | 2 | 218 | |||
AK301791.1 | - | 2 | 217 | |||
BC003183.1 | - | 2 | 234 | 2958852 | ||
BC004175.2 | - | 2 | 234 | 2958852 | ||
DB461868.1 | - | 2 | 284 | |||
DC399782.1 | - | 2 | 265 |