Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001323517.1 | NP_001310446.1 | 6 | 757 | ||
NM_001323518.1 | NP_001310447.1 | 6 | 609 | |||
NM_006744.3 | NP_006735.2 | 6 | 671 | |||
GenBank mRNA | BC020633.1 | - | 6 | 671 | 4767481 | |
BG565176.1 | - | 6 | 615 | |||
BI712834.1 | - | 1 | 277 | |||
BX495987.1 | - | 2 | 231 | |||
X00129.1 | - | 6 | 632 |