Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276261.1 | NP_001263190.1 | 9, 10 | 1226 | ||
NM_001282923.1 | NP_001269852.1 | 16, 17 | 1792 | |||
NM_001322214.1 | NP_001309143.1 | 9, 10 | 1226 | |||
NM_017790.4 | NP_060260.3 | 16, 17 | 1686 | |||
NM_130795.3 | NP_570613.2 | 9, 10 | 1226 | |||
NM_144488.5 | NP_652759.3 | 19, 20 | 2130 | |||
XM_006717219.2 | XP_006717282.1 | 18, 19 | 1976 | |||
XM_006717220.1 | XP_006717283.1 | 16, 17 | 1752 | |||
XM_006717222.1 | XP_006717285.1 | 9, 10 | 1102 | |||
XM_011518893.1 | XP_011517195.1 | 19, 20 | 2128 | |||
XM_017015004.1 | XP_016870493.1 | 15, 16 | 1500 | |||
XM_017015005.1 | XP_016870494.1 | 3, 4 | 339 | |||
GenBank mRNA | AF463495.1 | - | 9, 10 | 1226 | ||
AF490839.1 | - | 16, 17 | 1607 | |||
AF490840.1 | - | 9, 10 | 1076 | |||
AK000377.1 | - | 8, 9 | 1203 | |||
AK074977.1 | - | 16, 17 | 1656 | |||
AK096840.1 | - | 9, 10 | 1146 | |||
AK125059.1 | - | 16, 17 | 1763 | |||
AK128127.1 | - | 19, 20 | 2128 | |||
AK222888.1 | - | 8, 9 | 1203 | |||
BC042555.1 | - | 9, 10 | 1131 | 5175121 |