Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126121.1 | NP_001119593.1 | 4 | 480 | ||
NM_001126122.1 | NP_001119594.1 | 3 | 390 | |||
NM_021734.4 | NP_068380.3 | 4 | 464 | |||
XM_005257559.3 | XP_005257616.1 | 4 | 1102 | |||
XM_005257560.2 | XP_005257617.1 | 4 | 656 | |||
XM_005257561.3 | XP_005257618.1 | 4 | 992 | |||
XM_005257562.2 | XP_005257619.1 | 4 | 639 | |||
XM_006722007.2 | XP_006722070.1 | 4 | 512 | |||
XM_011525098.1 | XP_011523400.1 | 4 | 972 | |||
XM_017024926.1 | XP_016880415.1 | 3 | 1054 | |||
XM_017024927.1 | XP_016880416.1 | 2 | 728 | |||
XM_017024928.1 | XP_016880417.1 | 4 | 988 | |||
GenBank mRNA | AF182404.1 | - | 2 | 256 | ||
AJ251857.1 | - | 2 | 294 | |||
AK075239.1 | - | 5 | 486 | |||
AK097882.1 | - | 2 | 1166 | |||
AK225881.1 | - | 2 | 293 | |||
AL833387.1 | - | 4 | 431 | |||
AY346372.1 | - | 2 | 256 | |||
BC001075.2 | - | 4 | 392 | 2821142 | ||
BC005120.1 | - | 4 | 392 | 2821142 | ||
BU177306.1 | - | 3 | 349 | |||
DA605220.1 | - | 4 | 445 | |||
DB482052.1 | - | 2 | 220 |