Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031712.2 | NP_001026882.2 | 10 | 1090 | ||
XM_017011168.1 | XP_016866657.1 | 9 | 898 | |||
XM_017011169.1 | XP_016866658.1 | 9 | 1060 | |||
GenBank mRNA | AF182423.1 | - | 12 | 1221 | ||
AF532977.1 | - | 10 | 1055 | |||
BC056878.1 | - | 10 | 1090 | 4443778 |