Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282719.1 | NP_001269648.1 | 3 | 396 | ||
NM_001282720.1 | NP_001269649.1 | 3 | 378 | |||
NM_001282721.1 | NP_001269650.1 | 3 | 366 | |||
NM_001282722.1 | NP_001269651.1 | 2 | 222 | |||
NM_001282723.1 | NP_001269652.1 | 4 | 522 | |||
NM_001282724.1 | NP_001269653.1 | 3 | 396 | |||
NM_021925.3 | NP_068744.1 | 4 | 522 | |||
NR_104233.1 | 3 | 366 | ||||
XM_011533014.2 | XP_011531316.1 | 4 | 549 | |||
GenBank mRNA | AK025473.1 | - | 4 | 460 | ||
AK299244.1 | - | 4 | 463 | |||
AK299555.1 | - | 3 | 367 | |||
AK300494.1 | - | 3 | 318 | |||
AK301493.1 | - | 3 | 367 | |||
AK316162.1 | - | 3 | 325 | |||
AK316204.1 | - | 2 | 193 | |||
AK316310.1 | - | 3 | 349 | |||
BC017473.1 | - | 4 | 479 | 3611020 | ||
DC421670.1 | - | 3 | 378 |