Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C14orf93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130706.2 | NP_001124178.1 | 3 | 466 | ||
NM_001130708.2 | NP_001124180.1 | 2 | 387 | |||
NM_001282970.1 | NP_001269899.1 | 2 | 633 | |||
NM_021944.3 | NP_068763.2 | 2 | 633 | |||
XM_005267971.1 | XP_005268028.1 | 2 | 535 | |||
XM_005267972.1 | XP_005268029.1 | 2 | 436 | |||
XM_011537059.1 | XP_011535361.1 | 2 | 619 | |||
XM_011537060.2 | XP_011535362.1 | 3 | 431 | |||
XM_011537062.1 | XP_011535364.1 | 2 | 1688 | |||
XM_017021569.1 | XP_016877058.1 | 3 | 4498 | |||
XM_017021570.1 | XP_016877059.1 | 3 | 3783 | |||
XM_017021571.1 | XP_016877060.1 | 4 | 3904 | |||
XM_017021572.1 | XP_016877061.1 | 3 | 3743 | |||
XM_017021573.1 | XP_016877062.1 | 3 | 640 | |||
XM_017021574.1 | XP_016877063.1 | 4 | 3698 | |||
GenBank mRNA | AK022216.1 | - | 2 | 575 | ||
AK023026.1 | - | 2 | 320 | |||
BC014299.2 | - | 2 | 552 | 3936989 | ||
BX161383.1 | - | 4 | 570 | |||
BX161417.1 | - | 3 | 393 | |||
DA891690.1 | - | 2 | 387 |