Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001192.2 | NP_001183.2 | 1 | 291 | ||
GenBank mRNA | AY684975.1 | - | 1 | 175 | ||
BC058291.1 | - | 1 | 214 | 6736094 | ||
EF152353.1 | - | 1 | 70 | |||
EF152355.1 | - | 1 | 185 | |||
Z14954.1 | - | 1 | 139 | |||
Z29575.1 | - | 1 | 291 |