Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000983.3 | NP_000974.1 | 3 | 187 | ||
GenBank mRNA | AK097075.1 | - | 3 | 163 | ||
AK311749.1 | - | 3 | 161 | |||
BC035566.1 | - | 3 | 187 | 5109124 | ||
BC058887.1 | - | 3 | 145 | 6141451 | ||
BC066314.1 | - | 3 | 163 | 5303658 | ||
BU927988.1 | - | 201 | ||||
CA336134.1 | - | 3 | 190 | |||
CR456873.1 | - | 3 | 141 | |||
D17652.1 | - | 3 | 163 | |||
X59357.1 | - | 2 | 192 |