Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERINC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258031.1 | NP_001244960.1 | 9 | 1195 | ||
NM_001258032.1 | NP_001244961.1 | 8 | 1024 | |||
GenBank mRNA | AK097682.1 | - | 7 | 805 | ||
BC136669.1 | - | 6 | 820 | 9020659 | ||
BX647438.1 | - | 8 | 1024 | |||
DQ103711.1 | - | 8 | 984 |
Gene Symbol: SERF2-C15ORF63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF370428.1 | - | 1 | 67 |