Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025.4 | NP_001016.1 | 2 | 153 | ||
GenBank mRNA | AK291132.1 | - | 2 | 91 | ||
AK311772.1 | - | 2 | 91 | |||
BC070221.1 | - | 2 | 97 | 4770703 | ||
BG567497.1 | - | 2 | 92 | |||
BU594771.1 | - | 2 | 153 | |||
CF129812.1 | - | 2 | 242 | |||
CR456996.1 | - | 1 | 60 | |||
D14530.1 | - | 2 | 73 |