Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025.4 | NP_001016.1 | 2 | 109 | ||
GenBank mRNA | AK291132.1 | - | 2 | 47 | ||
AK311772.1 | - | 2 | 47 | |||
BC070221.1 | - | 2 | 53 | 4770703 | ||
BG567497.1 | - | 2 | 48 | |||
BU594771.1 | - | 2 | 109 | |||
CF129812.1 | - | 2 | 198 | |||
CR456996.1 | - | 1 | 16 | |||
D14530.1 | - | 2 | 29 |