Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128850.1 | NP_001122322.1 | 4 | 684 | ||
NM_004165.2 | NP_004156.1 | 4 | 675 | |||
GenBank mRNA | AK290613.1 | - | 4 | 674 | ||
BC011645.2 | - | 4 | 666 | 4299345 | ||
BC057815.1 | - | 4 | 683 | 30341066 | ||
L24564.1 | - | 4 | 655 |