Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000330.3 | NP_000321.1 | 4 | 273 | ||
GenBank mRNA | AF014459.1 | - | 4 | 253 | ||
AL049684.1 | - | 3 | 729 | |||
BQ636812.1 | - | 4 | 279 |