Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005619.4 | NP_005610.1 | 8 | 1658 | ||
NM_206900.2 | NP_996783.1 | 7 | 1439 | |||
NM_206901.2 | NP_996784.1 | 4 | 481 | |||
GenBank mRNA | AF004222.1 | - | 8 | 1557 | ||
AF004223.1 | - | 5 | 707 | |||
AF004224.1 | - | 4 | 449 | |||
AF038540.1 | - | 5 | 888 | |||
AK291967.1 | - | 7 | 1420 | |||
AK314158.1 | - | 7 | 1422 | |||
BC073874.1 | - | 8 | 1600 | 5242827 | ||
DA898294.1 | - | 4 | 481 |