Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RXRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006917.4 | NP_008848.1 | 2, 3 | 767 | ||
GenBank mRNA | AB307706.1 | - | 2, 3 | 305 | ||
AB593015.1 | - | 2, 3 | 411 | |||
AK312922.1 | - | 2, 3 | 697 | |||
AL538345.3 | - | 2, 3 | 605 | |||
BC012063.1 | - | 2, 3 | 493 | 4635470 | ||
CR456705.1 | - | 2, 3 | 305 | |||
HQ692845.1 | - | 2, 3 | 305 | |||
U38480.1 | - | 2, 3 | 332 |