Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005620.1 | NP_005611.1 | 3 | 299 | ||
GenBank mRNA | AK312184.1 | - | 3 | 234 | ||
BC001410.2 | - | 3 | 259 | 3140092 | ||
BC014354.1 | - | 3 | 241 | 3685742 | ||
BT009912.1 | - | 2 | 179 | |||
D38583.1 | - | 3 | 299 | |||
D49355.1 | - | 3 | 266 | |||
D50374.1 | - | 3 | 195 | |||
EU794659.1 | - | 3 | 256 | |||
X80201.1 | - | 3 | 256 |