Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303252.1 | NP_001290181.1 | 8, 9 | 990 | ||
NM_002969.4 | NP_002960.2 | 9, 10 | 1020 | |||
GenBank mRNA | AK098058.1 | - | 2, 3 | 1093 | ||
AK123665.1 | - | 2, 3 | 683 | |||
AK289770.1 | - | 4, 5 | 839 | |||
AK294422.1 | - | 6, 7 | 510 | |||
AK307421.1 | - | 6, 7 | 926 | |||
AK307724.1 | - | 2, 3 | 147 | |||
BC015741.1 | - | 9, 10 | 921 | 4907996 | ||
BX640648.1 | - | 7, 8 | 627 | |||
CR456515.1 | - | 8, 9 | 775 | |||
X79483.1 | - | 9, 10 | 803 | |||
Y10487.1 | - | 9, 10 | 976 |