Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017369.2 | NP_001017369.1 | 5 | 676 | ||
NM_006745.4 | NP_006736.1 | 6 | 962 | |||
XM_005263176.2 | XP_005263233.1 | 6 | 1079 | |||
GenBank mRNA | AK295432.1 | - | 5 | 622 | ||
BC010653.1 | - | 5 | 856 | 3866053 | ||
BC107879.1 | - | 6 | 924 | 6498540 | ||
U60205.1 | - | 5 | 858 | |||
U93162.1 | - | 6 | 922 |