Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005063.4 | NP_005054.3 | 1, 2 | 519 | ||
GenBank mRNA | AB032261.1 | - | 1, 2 | 409 | ||
AB208982.1 | - | 1, 2 | 276 | |||
AF097514.1 | - | 1, 2 | 264 | |||
AK222862.1 | - | 1, 2 | 236 | |||
AK312312.1 | - | 1, 2 | 301 | |||
BC005807.2 | - | 1, 2 | 206 | |||
BC062303.1 | - | 1, 2 | 405 | 6586153 | ||
BI827092.1 | - | 1, 2 | 520 | |||
BM546583.1 | - | 457 | ||||
Y13647.1 | - | 1, 2 | 237 |