Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001711.5 | NP_001702.1 | 2, 3 | 480 | ||
XM_017029724.1 | XP_016885213.1 | 1, 2 | 259 | |||
GenBank mRNA | AK092954.1 | - | 2, 3 | 388 | ||
AK094059.1 | - | 3, 4 | 289 | |||
AK222484.1 | - | 2, 3 | 389 | |||
AK223302.1 | - | 2, 3 | 360 | |||
AK291955.1 | - | 2, 3 | 391 | |||
AK293268.1 | - | 2, 3 | 416 | |||
AK293289.1 | - | 2, 3 | 315 | |||
AK297966.1 | - | 2, 3 | 389 | |||
AK298756.1 | - | 1, 2 | 189 | |||
AU142113.1 | - | 2, 3 | 416 | |||
BC002416.2 | - | 2, 3 | 373 | 3162633 | ||
BC004244.1 | - | 2, 3 | 383 | 3503374 | ||
BT007323.1 | - | 1, 2 | 244 | |||
DC299956.1 | - | 2, 3 | 480 | |||
DC419932.1 | - | 2, 3 | 460 | |||
DC425011.1 | - | 2, 3 | 441 | |||
J04599.1 | - | 2, 3 | 364 |