Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007099.2 | NP_001007100.1 | 4, 5 | 414 | ||
NM_001007100.2 | NP_001007101.1 | 4, 5 | 405 | |||
NM_001007250.2 | NP_001007251.1 | 3, 4 | 284 | |||
NM_001193599.1 | NP_001180528.1 | 14, 15 | 1639 | |||
NM_001193600.1 | NP_001180529.1 | 14, 15 | 1579 | |||
NM_001193617.1 | NP_001180546.1 | 14, 15 | 1653 | |||
NM_002979.4 | NP_002970.2 | 15, 16 | 1711 | |||
XM_005271103.4 | XP_005271160.1 | 14, 15 | 1524 | |||
XM_017002046.1 | XP_016857535.1 | 5, 6 | 979 | |||
GenBank mRNA | AB208789.1 | - | 16, 17 | 4406 | ||
AK294631.1 | - | 14, 15 | 1582 | |||
AK295214.1 | - | 14, 15 | 1596 | |||
AK308105.1 | - | 14, 15 | 1521 | |||
AK312856.1 | - | 15, 16 | 1662 | |||
BC005911.1 | - | 4, 5 | 404 | |||
CA867670.1 | - | 3, 4 | 241 | |||
DB459183.1 | - | 4, 5 | 383 | |||
M55421.1 | - | 4, 5 | 414 | |||
M75883.1 | - | 15, 16 | 1564 | |||
M75884.1 | - | 7, 8 | 850 | |||
S52450.1 | - | 4, 5 | 414 |