Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002982.3 | NP_002973.1 | 2 | 219 | ||
GenBank mRNA | AK311960.1 | - | 2 | 211 | ||
AV733621.2 | - | 2 | 219 | |||
BC009716.1 | - | 2 | 202 | 3901489 | ||
BG530064.1 | - | 2 | 232 | |||
BT007329.1 | - | 2 | 146 | |||
M24545.1 | - | 2 | 215 | |||
M26683.1 | - | 1 | 84 | |||
M28226.1 | - | 2 | 192 | |||
S69738.1 | - | 2 | 177 | |||
S71513.1 | - | 2 | 199 | |||
X14768.1 | - | 2 | 199 |