Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002982.3 | NP_002973.1 | 3 | 356 | ||
GenBank mRNA | AK311960.1 | - | 3 | 348 | ||
AV733621.2 | - | 3 | 356 | |||
BC009716.1 | - | 3 | 339 | 3901489 | ||
BG530064.1 | - | 3 | 369 | |||
BT007329.1 | - | 3 | 283 | |||
M24545.1 | - | 3 | 352 | |||
M26683.1 | - | 1 | 601 | |||
M28226.1 | - | 3 | 329 | |||
S69738.1 | - | 3 | 314 | |||
S71513.1 | - | 3 | 336 | |||
X14768.1 | - | 3 | 336 |