Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2O | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022066.3 | NP_071349.3 | 13 | 2146 | ||
XM_005257567.1 | XP_005257624.1 | 13 | 2133 | |||
XM_017024934.1 | XP_016880423.1 | 13 | 2058 | |||
XM_017024935.1 | XP_016880424.1 | 13 | 2055 | |||
GenBank mRNA | AB051521.1 | - | 13 | 2146 | ||
AK025999.1 | - | 5 | 625 | |||
AK225412.1 | - | 5 | 625 | |||
AL832432.1 | - | 6 | 1071 | |||
BC036820.1 | - | 8 | 1282 | |||
BC051868.2 | - | 5 | 489 | 6162673 |