Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC17A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302643.1 | NP_001289572.1 | 11 | 1255 | ||
NM_022082.3 | NP_071365.3 | 10 | 1193 | |||
XM_011528978.2 | XP_011527280.1 | 10 | 1105 | |||
GenBank mRNA | AK027065.1 | - | 3 | 182 | ||
AK074107.1 | - | 9 | 1989 | |||
AK095473.1 | - | 10 | 1044 | |||
AK125597.1 | - | 10 | 1716 | |||
AK298503.1 | - | 10 | 1099 | |||
BC025312.1 | - | 11 | 1195 | 4563089 | ||
BC027447.1 | - | 6 | 414 | |||
BC038593.1 | - | 10 | 1035 |