Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318253.1 | NP_001305182.1 | 14 | 1776 | ||
NM_133171.4 | NP_573403.1 | 14 | 1708 | |||
NM_182764.2 | NP_877496.1 | 13 | 1604 | |||
XM_005260496.3 | XP_005260553.1 | 14 | 1273 | |||
XM_005260498.3 | XP_005260555.1 | 13 | 1635 | |||
XM_005260499.3 | XP_005260556.1 | 13 | 1606 | |||
XM_005260500.3 | XP_005260557.1 | 12 | 1134 | |||
XM_005260501.4 | XP_005260558.1 | 13 | 1144 | |||
XM_006723854.3 | XP_006723917.1 | 13 | 1207 | |||
XM_017028009.1 | XP_016883498.1 | 13 | 1579 | |||
XM_017028010.1 | XP_016883499.1 | 12 | 1082 | |||
XM_017028011.1 | XP_016883500.1 | 12 | 1535 | |||
GenBank mRNA | AB058737.1 | - | 13 | 1219 | ||
AF398886.1 | - | 12 | 1147 | |||
AF417861.1 | - | 12 | 1079 | |||
AK021718.1 | - | 3 | 275 | |||
AK023103.1 | - | 2 | 35 | |||
AK293861.1 | - | 12 | 1172 | |||
AK299318.1 | - | 16 | 1319 | |||
AK302710.1 | - | 4 | 511 | |||
AL713765.1 | - | 3 | 253 | |||
BC000143.4 | - | 13 | 1284 | 3047992 | ||
BC021079.2 | - | 3 | 271 | |||
BX648819.1 | - | 13 | 1185 |