Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022095.3 | NP_071378.1 | 21 | 3368 | ||
XM_005260504.4 | XP_005260561.1 | 21 | 4081 | |||
XM_017028012.1 | XP_016883501.1 | 20 | 3139 | |||
GenBank mRNA | AF395833.1 | - | 21 | 3345 | ||
AK026157.1 | - | 13 | 1883 | |||
AK074214.1 | - | 6 | 1003 | |||
AK122963.1 | - | 4 | 1559 | |||
AK296989.1 | - | 18 | 2939 | |||
BC041331.2 | - | 18 | 2583 | |||
BC142981.1 | - | 18 | 2583 |