Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022100.2 | NP_071383.1 | 3 | 420 | ||
NR_037606.1 | 3 | 512 | ||||
GenBank mRNA | AK310915.1 | - | 3 | 482 | ||
AK311759.1 | - | 3 | 390 | |||
AL049705.1 | - | 3 | 390 | |||
BC009788.1 | - | 3 | 386 | 4132323 | ||
CB957578.1 | - | 3 | 423 |