Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXorf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170569.1 | NP_001164040.1 | 7 | 811 | ||
NM_001170570.1 | NP_001164041.1 | 6 | 687 | |||
NM_022101.3 | NP_071384.1 | 7 | 729 | |||
GenBank mRNA | AK096297.1 | - | 7 | 775 | ||
AK298877.1 | - | 6 | 665 | |||
BC023506.2 | - | 7 | 690 | |||
BQ223940.1 | - | 7 | 694 |
Gene Symbol: LOC728024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003671.2 | 1 | 692 | |||
GenBank mRNA | CR977513.1 | - | 1 | 599 |