Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321355.1 | NP_001308284.1 | 6 | 1235 | ||
NM_001321356.1 | NP_001308285.1 | 7 | 1736 | |||
NM_022103.3 | NP_071386.3 | 5 | 1134 | |||
XM_011527208.1 | XP_011525510.1 | 6 | 3380 | |||
XM_011527209.1 | XP_011525511.1 | 5 | 2894 | |||
GenBank mRNA | AK024073.1 | - | 1 | 497 | ||
AK056508.1 | - | 5 | 1076 | |||
AK096513.1 | - | 7 | 1665 | |||
AK126957.1 | - | 6 | 2526 | |||
BC074898.2 | - | 1 | 332 | 30915250 | ||
BC074899.2 | - | 1 | 332 | 30915479 | ||
BC136388.1 | - | 4 | 970 | 9020375 | ||
BC150593.1 | - | 4 | 970 | 9056963 | ||
BX647401.1 | - | 5 | 1107 | |||
CR936605.1 | - | 6 | 1200 |