Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXCT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022120.1 | NP_071403.1 | 1 | 890 | ||
GenBank mRNA | AB050193.2 | - | 1 | 816 | ||
AK093089.1 | - | 1 | 864 | |||
AK314586.1 | - | 1 | 797 | |||
AY013700.1 | - | 1 | 890 | |||
BC030015.1 | - | 1 | 809 |