Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287580.1 | NP_001274509.1 | 6 | 1620 | ||
NM_022128.2 | NP_071411.1 | 5 | 501 | |||
GenBank mRNA | AJ404857.1 | - | 5 | 461 | ||
AK300989.1 | - | 5 | 455 | |||
AY643715.1 | - | 5 | 493 | |||
BC017425.1 | - | 5 | 503 | 4694872 | ||
BX648909.1 | - | 6 | 1591 |