Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022133.3 | NP_071416.2 | 3 | 810 | ||
NM_152836.2 | NP_690049.1 | 2 | 564 | |||
NM_152837.2 | NP_690050.1 | 2 | 564 | |||
XM_005251282.4 | XP_005251339.1 | 3 | 517 | |||
XM_005251283.2 | XP_005251340.1 | 4 | 654 | |||
GenBank mRNA | AF305779.1 | - | 2 | 415 | ||
AK290903.1 | - | 2 | 480 | |||
AL833763.1 | - | 2 | 471 | |||
BC033630.1 | - | 2 | 472 | 5224216 | ||
BG739930.1 | - | 2 | 478 | |||
CB962009.1 | - | 2 | 542 |