Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308419.1 | NP_001295348.1 | 2 | 169 | ||
NM_022165.2 | NP_071448.1 | 2 | 169 | |||
XM_006723323.3 | XP_006723386.1 | 2 | 189 | |||
XM_017027131.1 | XP_016882620.1 | 2 | 191 | |||
GenBank mRNA | AF311862.1 | - | 2 | 147 | ||
AY358744.1 | - | 2 | 169 | |||
BC027618.2 | - | 2 | 143 | 4830017 | ||
BM069859.1 | - | 2 | 126 |