Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIN7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308419.1 | NP_001295348.1 | 2, 3 | 206 | ||
NM_022165.2 | NP_071448.1 | 2, 3 | 206 | |||
GenBank mRNA | AF311862.1 | - | 2, 3 | 184 | ||
AY358744.1 | - | 2, 3 | 206 | |||
BC027618.2 | - | 2, 3 | 180 | 4830017 | ||
BM069859.1 | - | 2, 3 | 163 |