Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022346.4 | NP_071741.2 | 15 | 2453 | ||
NR_073124.1 | 14 | 2343 | ||||
XM_011513876.2 | XP_011512178.1 | 14 | 2252 | |||
XM_017008543.1 | XP_016864032.1 | 15 | 2343 | |||
XM_017008544.1 | XP_016864033.1 | 14 | 2208 | |||
GenBank mRNA | AB013299.1 | - | 15 | 2327 | ||
AF235023.1 | - | 15 | 2256 | |||
AF331796.1 | - | 15 | 2364 | |||
AK022512.1 | - | 15 | 2284 | |||
AK023147.1 | - | 14 | 2034 | |||
AK027511.1 | - | 6 | 790 | |||
BC000827.1 | - | 4 | 541 | |||
BC068467.1 | - | 15 | 2270 | 4821684 | ||
BC101476.1 | - | 15 | 2241 | 8068982 |