Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042595.2 | NP_001036060.1 | 2 | 101 | ||
NM_001098821.1 | NP_001092291.1 | 2 | 363 | |||
NM_001098822.1 | NP_001092292.1 | 2 | 363 | |||
NM_001098823.1 | NP_001092293.1 | 2 | 363 | |||
NM_001098824.1 | NP_001092294.1 | 2 | 363 | |||
NM_001098825.1 | NP_001092295.1 | 2 | 171 | |||
GenBank mRNA | AK130820.1 | - | 2 | 672 | ||
AL833095.1 | - | 2 | 333 | |||
BC063705.1 | - | 2 | 73 | 4804092 | ||
BF684543.1 | - | 2 | 163 | |||
BG697182.1 | - | 2 | 101 | |||
BM726200.1 | - | 2 | 138 | |||
BM931402.1 | - | 2 | 318 | |||
BX090160.1 | - | 1 | 46 | |||
DA673235.1 | - | 2 | 363 | |||
R01772.1 | - | 170 | ||||
R67551.1 | - | 1 | 48 |