Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP2K4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281435.1 | NP_001268364.1 | 11, 12 | 1176 | ||
NM_003010.3 | NP_003001.1 | 10, 11 | 1143 | |||
XM_005256753.2 | XP_005256810.1 | 9, 10 | 982 | |||
XM_005256755.2 | XP_005256812.1 | 9, 10 | 964 | |||
XM_011523977.2 | XP_011522279.1 | 8, 9 | 854 | |||
GenBank mRNA | AK122997.1 | - | 3, 4 | 1255 | ||
AK131544.1 | - | 11, 12 | 1161 | |||
AK294923.1 | - | 11, 12 | 1019 | |||
AK313053.1 | - | 10, 11 | 1096 | |||
AK316100.1 | - | 9, 10 | 910 | |||
AK316177.1 | - | 8, 9 | 745 | |||
BC036032.1 | - | 10, 11 | 1143 | 5272439 | ||
BT019676.1 | - | 10, 11 | 1074 | |||
CR536564.1 | - | 10, 11 | 1074 | |||
L36870.1 | - | 10, 11 | 1083 | |||
U17743.1 | - | 10, 11 | 1094 |