Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETMAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243723.1 | NP_001230652.1 | 1 | 105 | ||
NM_001276325.1 | NP_001263254.1 | 1 | 105 | |||
NM_001320676.1 | NP_001307605.1 | 1 | 105 | |||
NM_001320677.1 | NP_001307606.1 | 1 | 105 | |||
NM_001320678.1 | NP_001307607.1 | 1 | 105 | |||
NM_006515.3 | NP_006506.3 | 1 | 105 | |||
GenBank mRNA | AF054989.1 | - | 1 | 31 | ||
AK222734.1 | - | 1 | 41 | |||
AK297865.1 | - | 1 | 40 | |||
AK302296.1 | - | 1 | 39 | |||
AK307537.1 | - | 1 | 105 | |||
BC008931.2 | - | 1 | 24 | |||
BC011635.2 | - | 1 | 31 | 4080032 | ||
DC357038.1 | - | 1 | 105 | |||
U80776.1 | - | 1 | 22 |